Merck
Drug Studies on Rett Syndrome: From Bench to Bedside.
Journal of autism and developmental disorders (2020-02-06)
Mohan Gomathi, Subramanian Padmapriya, Vellingiri Balachandar
ABSTRACT
Drug studies on Rett syndrome (RTT) have drastically increased over the past few decades. This review aims to provide master data on bench-to-bedside drug studies involving RTT. A comprehensive literature review was performed by searching in PUBMED, MEDLINE and Google Scholar, international, national and regional clinical trial registries and pharmaceutical companies using the keywords "Rett syndrome treatment and/or drug or compound or molecule". Seventy drugs were investigated in non-clinical (N = 65 animal/cell line-based studies; N = 5 iPSC-based study) and clinical trials (N = 34) for ameliorating the symptoms of RTT. Though there is good progress in both clinical and non-clinical studies, none of these drugs entered phase III/IV for being launched as a therapeutic agent for RTT.
Sigma-Aldrich
Glycerol trienanthate, ≥94%