Merck
  • Septal Flash-like Motion of the Earlier Activated Ventricular Wall Represents the Pathophysiology of Mechanical Dyssynchrony in Single-Ventricle Anatomy.
Septal Flash-like Motion of the Earlier Activated Ventricular Wall Represents the Pathophysiology of Mechanical Dyssynchrony in Single-Ventricle Anatomy.
Journal of the American Society of Echocardiography : official publication of the American Society of Echocardiography (2020-02-25)
Yohsuke Hayama, Aya Miyazaki, Hideo Ohuchi, Hikari Miike, Jun Negishi, Heima Sakaguchi, Kenichi Kurosaki, Shuji Shimizu, Toru Kawada, Masaru Sugimachi
ABSTRACT
In biventricular physiology, abnormal septal motion is a hallmark of mechanical dyssynchrony in the left bundle branch block. However, in single-ventricle (SV) physiology, morphologic variations in systemic ventricles pose a challenge in evaluating the negative impact of mechanical dyssynchrony. The present study aimed to characterize the pathologic dyssynchronous contraction patterns in patients with SV. In this retrospective study, 70 consecutive postoperative patients with SV anatomy with prolonged QRS duration (25 female patients; median age, 14 years) were enrolled. We divided each SV into two regions and analyzed independent strains using two-dimensional speckle-tracking echocardiography. From an earlier activated ventricular wall, we calculated the strain ratio (Rstrains) of two values (%) during the QRS period and the ejection period: (100 + Strainejection)/(100 + StrainQRS). We reviewed the clinical profiles, B-type natriuretic peptide plasma levels, exercise capacity, and morbidity. Six patients who underwent cardiac resynchronization therapy (CRT) were analyzed regarding changes in strain patterns and ventricular volume. Higher Rstrains, indicating a preceding contraction and subsequent dyskinetic dilation of the earlier activated ventricular wall, was associated with increased B-type natriuretic peptide, reduced exercise capacity, and poor outcome. However, delayed contraction of the later activated ventricular wall was not associated with the effects. Decreases in Rstrains and ventricular volume reductions were observed in all patients after CRT. A specific strain pattern in an earlier activated ventricular wall indicates mechanical dyssynchrony in patients with SV. This pattern is very similar to the septal flash in adult patients with left bundle branch block. This strategy might be a promising approach for selecting appropriate candidates for CRT in patients with SV.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%