Merck
  • Intravitreal bromfenac liposomal suspension (100 μg / 0.1 ml). A safety study in rabbit eyes.
Intravitreal bromfenac liposomal suspension (100 μg / 0.1 ml). A safety study in rabbit eyes.
Experimental eye research (2020-03-27)
Idaira Sánchez-Santos, Gustavo A García-Sánchez, Roberto Gonzalez-Salinas, Mónica Anayatzin Linares-Alba, Abelardo A Rodríguez-Reyes, Rodrigo García-Santisteban, Vanessa Tirado-González, Elsa Hernández-Piñamora, Daniel García-Arzate, Virgilio Morales-Cantón, Hugo Quiroz-Mercado
ABSTRACT
There is a need to find alternative treatments for MEe. Bromfenac has shown promise in inhibiting the COX-2 enzymatic pathway that partially causes the inflammatory cascade which contributes to the precipitation of ME. However, like other NSAID's, its intraocular half-life is limited. We hypothesize that a delayed-release liposome formulation containing bromfenac might provide a similar anti-inflammatory effect as long-lasting steroid release systems without the well-known steroidal side-effects. We introduced a novel formulation with these characteristics into the vitreous cavity of rabbit eyes in order to evaluate its safety profile. 10 left eyes of rabbits were injected with the liposome-encapsulated bromfenac suspension (100 μg/0.1 ml). Basal ERG's were recorded. Total follow-up time was 3 months, at which point ERG's were repeated and eyes were enucleated for histopathological study. Total amplitude and implicit times were recorded. A difference of 25% in either recording was considered significant. Significance was assessed using the paired-t test and Wilcoxon matched-pairs signed-rank test. A p-value of <0.05 was considered significant. No significant changes were recorded in ERG measurements after 3 months when compared to basal measurements. Histopathological analysis of retinal specimens found no traces of liposome-induced toxicity. The liposome-encapsulated bromfenac suspension (100 μg/0.1 ml) is not toxic and has been proven safe to use in an animal model. Therefore, this formulation shows promise as a possible future alternative treatment for ME and should be further studied to show its biological effect and efficacy.
Sigma-Aldrich
[2-Amino-3-(4-bromobenzoyl)phenyl]acetic acid