Merck
  • Targeted mutation of secretogranin-2 disrupts sexual behavior and reproduction in zebrafish.
Targeted mutation of secretogranin-2 disrupts sexual behavior and reproduction in zebrafish.
Proceedings of the National Academy of Sciences of the United States of America (2020-05-30)
Kimberly Mitchell, Wo Su Zhang, Chunyu Lu, Binbin Tao, Lu Chen, Wei Hu, Vance L Trudeau
ABSTRACT
The luteinizing hormone surge is essential for fertility as it triggers ovulation in females and sperm release in males. We previously reported that secretoneurin-a, a neuropeptide derived from the processing of secretogranin-2a (Scg2a), stimulates luteinizing hormone release, suggesting a role in reproduction. Here we provide evidence that mutation of the scg2a and scg2b genes using TALENs in zebrafish reduces sexual behavior, ovulation, oviposition, and fertility. Large-scale spawning within-line crossings (n = 82 to 101) were conducted. Wild-type (WT) males paired with WT females successfully spawned in 62% of the breeding trials. Spawning success was reduced to 37% (P = 0.006), 44% (P = 0.0169), and 6% (P < 0.0001) for scg2a -/- , scg2b -/- , and scg2a -/- ;scg2b -/- mutants, respectively. Comprehensive video analysis indicates that scg2a -/- ;scg2b -/- mutation reduces all male courtship behaviors. Spawning success was 47% in saline-injected WT controls compared to 11% in saline-injected scg2a -/- ;scg2b -/- double mutants. For these mutants, spawning success increased 3-fold following a single intraperitoneal (i.p.) injection of synthetic secretoneurin-a (P = 0.0403) and increased 3.5-fold with injection of human chorionic gonadotropin (hCG). Embryonic survival at 24 h remained on average lower in scg2a -/- ;scg2b -/- fish compared to WT injected with secretoneurin-a (P < 0.001). Significant reductions in the expression of gonadotropin-releasing hormone 3 in the hypothalamus, and luteinizing hormone beta and glycoprotein alpha subunits in the pituitary provide evidence for disrupted hypothalamo-pituitary function in scg2a and scg2b mutant fish. Our results indicate that secretogranin-2 is required for optimal reproductive function and support the hypothesis that secretoneurin is a reproductive hormone.
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
Chorionic Gonadotropin, Human Urine, Standard Grade, Chorionic Gonadotropin, Human Urine, Standard Grade, CAS 9002-61-3, is a native glycoprotein hormone that is synthesized by chorionic tissue of the placenta and found in urine during pregnancy.