Merck
  • α-Methyltyrosine, a tyrosine hydroxylase inhibitor, decreases stress response in zebrafish (Danio rerio).
α-Methyltyrosine, a tyrosine hydroxylase inhibitor, decreases stress response in zebrafish (Danio rerio).
General and comparative endocrinology (2017-07-19)
Renan Idalencio, Heloísa Helena de Alcântara Barcellos, Fabiana Kalichak, João Gabriel Santos da Rosa, Thiago Acosta Oliveira, Murilo Sander de Abreu, Michele Fagundes, Fernanda Dametto, Letícia Marcheto, Caio Maximino de Oliveira, Leonardo José Gil Barcellos
ABSTRACT
In this article, we show that the tyrosine hydroxylase inhibitor α-Methyl-l-tyrosine (AMPT) decreased the responsiveness of the zebrafish stress axis to an acute stressful challenge. These effects were specific for responses to stimulation, since unstimulated (basal) cortisol levels were not altered by AMPT. Moreover, AMPT decreased the stress response 15min after stimulation, but not after that time period. To our knowledge, this is the first report about the effects of AMPT on the neuroendocrine axis of adult zebrafish in acute stress responses. Overall, these results suggest a mechanism of catecholamine-glucocorticoid interplay in neuroendocrine responses of fish, pointing an interesting avenue for physiological research, as well as an important endpoint that can be disrupted by environmental contamination. Further experiments will unravel the mechanisms by which AMPT blocked the cortisol response.
Sigma-Aldrich
α-Methyl-L-tyrosine, ≥98% (TLC)