Merck
  • Cloning and sequencing of a dextranase-encoding cDNA from Penicillium minioluteum.
Cloning and sequencing of a dextranase-encoding cDNA from Penicillium minioluteum.
FEMS microbiology letters (1996-10-01)
B Garcia, E Margolles, H Roca, D Mateu, M Raices, M E Gonzales, L Herrera, J Delgado
ABSTRACT
A cDNA from Penicillium minioluteum HI-4 encoding a dextranase (1,6-alpha-glucan hydrolase, EC 3.2.1.11) was isolated and characterized. cDNA clones corresponding to genes expressed in dextran-induced cultures were identified by differential hybridization. Southern hybridization and restriction mapping analysis of selected clones revealed four different groups of cDNAs. The dextranase cDNA was identified after expressing a cDNA fragment from each of the isolated groups of cDNA clones in the Escherichia coli T7 system. The expression of a 2 kb cDNA fragment in E. coli led to the production of a 67 kDa protein which was recognized by an anti-dextranase polyclonal antibody. The cDNA contains 2109 bp plus a poly(A) tail, coding for a protein of 608 amino acids, including 20 N-terminal amino acid residues which might correspond to a signal peptide. There was 29% sequence identity between the P. minioluteum dextranase and the dextranase from Arthrobacter sp. CB-8.
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid
Pricing and availability is not currently available.