Merck
  • Impact of NHC ligand conformation and solvent concentration on the ruthenium-catalyzed ring-closing metathesis reaction.
Impact of NHC ligand conformation and solvent concentration on the ruthenium-catalyzed ring-closing metathesis reaction.
Journal of the American Chemical Society (2009-06-19)
Michele Gatti, Ludovic Vieille-Petit, Xinjun Luan, Ronaldo Mariz, Emma Drinkel, Anthony Linden, Reto Dorta
ABSTRACT
Two saturated N-heterocyclic carbene ligands with substituted naphthyl side chains were used for the preparation of Blechert-type ruthenium metathesis precatalysts. The resulting conformers of the complexes were separated and unambiguously assigned by X-ray diffraction studies. All new complexes were compared in terms of activity to the original, SIMes-derived Blechert catalyst and were shown to be superior. A study on the impact of solvent concentration in RCM reactions using the most active of these new catalysts ultimately led to the ring closing of a variety of substrates at very low catalyst loadings.
Sigma-Aldrich
Formaldehyde dibutyl acetal, puriss., ≥97.0% (GC)