Merck
  • Sensitive determination of RDX, nitroso-RDX metabolites, and other munitions in ground water by solid-phase extraction and isotope dilution liquid chromatography-atmospheric pressure electro-spray [correction of chemical] ionization mass spectrometry.
Sensitive determination of RDX, nitroso-RDX metabolites, and other munitions in ground water by solid-phase extraction and isotope dilution liquid chromatography-atmospheric pressure electro-spray [correction of chemical] ionization mass spectrometry.
Journal of chromatography. A (1999-07-10)
D A Cassada, S J Monson, D D Snow, R F Spalding
ABSTRACT
Recent improvements in the LC-MS interface have increased the sensitivity and selectivity of this instrument in the analysis of polar and thermally-labile aqueous constituents. Determination of RDX, nitroso-RDX metabolites, and other munitions was enhanced using LC-MS with solid-phase extraction, 15N3-RDX internal standard, and electrospray ionization (ESI) in negative ion mode. ESI produced a five-fold increase in detector response over atmospheric pressure chemical ionization (APCI) for the nitramine compounds, while the more energetic APCI produced more than twenty times the ESI response for nitroaromatics. Method detection limits in ESI for nitramines varied from 0.03 microgram l-1 for MNX to 0.05 microgram l-1 for RDX.
Product Description
Supelco
1,3-Dinitrobenzene, analytical standard
Pricing and availability is not currently available.