- 1,6-Carbene transfer: gold-catalyzed oxidative diyne cyclizations.
1,6-Carbene transfer: gold-catalyzed oxidative diyne cyclizations.
Journal of the American Chemical Society (2013-09-21)
Pascal Nösel, Laura Nunes dos Santos Comprido, Tobias Lauterbach, Matthias Rudolph, Frank Rominger, A Stephen K Hashmi
PMID24050384
ABSTRACT
In the presence of a gold catalyst an unprecedented oxidative cyclization of diynes takes place. The reaction cascade is initiated by an oxygen transfer from a N-oxide onto a gold-activated alkyne. The formed α-oxo carbene is transferred across the second alkyne yielding a stabilized vinyl carbene/cation. Alkyl migration or sp(3)-CH insertion then terminates the catalytic cycle by formation of highly substituted functionalized indenones. A 1,6-carbene shift could be supported by the oxidation of the vinyl carbene. This protocol represents an attractive alternative to procedures which are based on the metal-catalyzed decomposition of hazardous, not easily accessible, diazo compounds.
MATERIALS