Merck
  • A functional test identifies dopamine agonists selective for D3 versus D2 receptors.
A functional test identifies dopamine agonists selective for D3 versus D2 receptors.
Neuroreport (1995-01-26)
F Sautel, N Griffon, D Lévesque, C Pilon, J C Schwartz, P Sokoloff
ABSTRACT
The functional potency of a series of dopamine agonists for increasing mitogenesis, measured by incorporation of [3H]thymidine, was established in transfected cell lines expressing human D2 or D3 receptors. The functional selectivity of agonists markedly differs from their binding selectivity. (+)7-OH-DPAT, pramipexole, quinerolane and PD 128,907, the most D3 receptor-selective compounds in binding studies, were 7, 15, 21 and 54 times more potent, respectively, at the D3 than at the D2 receptor in the functional test. Bromocriptine displayed a 10-fold functional selectivity toward the D2 receptor. The known behavioural actions of D3 selective agonists support a role for the D3 receptor in motor inhibitions, which should be taken into account for the treatment of motor dysfunctions by dopamine agonists.
Sigma-Aldrich
(+)-PD 128,907 hydrochloride, >97%, solid