Merck
  • CCN5 inhibits proliferation and promotes apoptosis of oral squamous cell carcinoma cells.
CCN5 inhibits proliferation and promotes apoptosis of oral squamous cell carcinoma cells.
Cell biology international (2020-01-01)
Shiqun Sun, Zhi Cui, Tongtong Yan, Jian Wu, Zhihui Liu
ABSTRACT
Oral squamous cell carcinoma (OSCC) is a common cancer with poor prognosis and high mortality. The role of CCN5 has attracted a great focus on the regulation of cancer progression. However, the biological function and mechanism of CCN5 in OSCC are still not well elucidated. The current study was designed to determine the effects of CCN5 on OSCC cell proliferation and apoptosis using two OSCC cell lines. Further, LY294002, a PI3K/AKT antagonist, was employed to explore the mechanism underlying the effects of CCN5 in the regulation of OSCC. The results showed that overexpression of CCN5 in TSCCa cells significantly reduced viable cell number, arrested cell cycle, and suppressed cell-cycle regulators (cyclin D1, cyclin E, and CDK2). CCN5 overexpression increased the apoptotic ratio and Hoechst-positive cell number, and altered the apoptotic-related proteins (caspase-3/9, Bax, and Bcl-2). However, CCN5 silencing induced opposite effects on cell proliferation and apoptosis in Tca-8113 cells. In addition, we observed that CCN5 knockdown increased the expression levels of PI3K (p85α and p110α) and phosphorylated AKT at serine 473 (p-AKT Ser473) in Tca-8113 cells. Inhibiting PI3K/AKT signaling with LY294002 rescued the apoptotic process in CCN5-silenced OSCC cells. Finally, xenograft analysis showed that CCN5 represses tumorigenesis of OSCC cells. These findings together suggest that CCN5 functions as a tumor suppressor for OSCC cell development through inactivation of PI3K/AKT signaling pathway, providing a potential candidate for OSCC therapy.
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays