Merck
  • Elucidation of Selectivity for Silver Ion Based on Metal-Induced H-Type Aggregation of Fluorescent Receptor.
Elucidation of Selectivity for Silver Ion Based on Metal-Induced H-Type Aggregation of Fluorescent Receptor.
Journal of fluorescence (2020-02-23)
Taner Arslan, Merve Umutlu, Orhan Güney
ABSTRACT
In present work, a new substituted phthalonitrile derivative was prepared by the nucleophilic displacement reaction and then highly soluble zinc phthalocyanine (ZnPc) with four peripheral 1-hydroxyhexan-3-ylthio groups was synthesized by cyclotetramerization and characterized by FTIR, 1H and 13C NMRs spectroscopies, fluorescence and UV-vis measurements. The optical property and quantum yield of ZnPc were elucidated in mixed solvent of ethanol/water with varying compositions. The pH-dependent fluorescence and absorbance spectra of ZnPc in the absence and presence of Ag+ ions were obtained to elucidate the optimum pH value that is convenient for stable complex formation in predetermined mixture. A comparative study for recognition of Ag+ ion has been carried out to evaluate the effect of the solution parameters on selective sensing ability of ZnPc as a fluorescent receptor. Interference effect was investigated by spectrofluorometric titration based on fluorescence quenching of ZnPc upon addition of Ag+ ion in the presence of foreign metal ions. Stepwise complexation/decomplexation cycles with Na2S-titration by fluorescence quenching/enhancement were investigated to establish the reversible response rate and reusability of ZnPc toward Ag+ ions. Graphical Abstract Selective determination of silver ion based on H-type aggregation of zinc phthalocyanine.
Sigma-Aldrich
3-Nitrophthalonitrile, 99%
Pricing and availability is not currently available.