Merck
  • [Studies of oligosaccharide specificity of perch roe fucolectin, using gold labeled neoglycoproteins].
[Studies of oligosaccharide specificity of perch roe fucolectin, using gold labeled neoglycoproteins].
Prikladnaia biokhimiia i mikrobiologiia (2003-03-11)
V E Piskarev, O Iu Praĭzel', R P Evstigneeva, I A Iamskov
ABSTRACT
The specificity of perch (Perca fluviatilis) roe fucolectin was studied using the protein dot blot technique, followed by detection with colloidal gold-labeled neoglycoproteins bearing human milk polysaccharides. The strongest binding was noted with the H type 1 pentasaccharide lacto-N-fucopentaose (LNFP I, Fuc alpha 1-2 Gal beta 1-3 GlcNAc beta 1-3 Gal beta 1-4Glc); the interaction with the H type 6 trisaccharide 2'-fucosyllactose (2-FL, Fuc alpha 1-2 Gal beta 1-4 Glc) was weaker. Binding of the perch lectin to the Lewis antigens (associated with tumors and embryonic tissues) was also studied. It was found that the lectin weakly interacted with the hexasaccharide lacto-N-difucohexaose I, Fuc alpha 1-2 Gal beta 1-3[Fuc alpha 1-4]GlcNac beta 1-3 Gal beta 1-4 Glc), but not with Lea, Lec, Lex antigens. Thus, perch roe lectin exhibited pronounced differences in carbohydrate specificity from other fucolectins--a feature that may be used in structural studies and isolation of fucose-containing glycoconjugates.
Sigma-Aldrich
Benzyl 2-acetamido-2-deoxy-3-O-β-D-galactopyranosyl-α-D-galactopyranoside