Merck
N-Acetylheparin pretreatment reduces infarct size in the rabbit.
Pharmacology (1999-02-02)
J L Park, K S Kilgore, K B Naylor, E A Booth, K L Murphy, B R Lucchesi
ABSTRACT
The ability of the heparin derivative, N-acetylheparin (NHEP) to protect the heart from regional ischemia/reperfusion injury was examined in vivo. NHEP (2 mg/kg i.v.) or vehicle was administered 2 h before occlusion of the left circumflex coronary (LCX) artery. Open-chest, anesthetized rabbits were subjected to 30 min of regional myocardial ischemia followed by 5 h of reperfusion. Myocardial myeloperoxidase activity, membrane attack complex (MAC) deposition and IL-8 generation were assessed in supernatant samples from the area at risk. Infarct size in rabbits pretreated with NHEP (32.5 +/- 3.8%, n = 10) decreased by 41% compared to infarct size in rabbits that received vehicle (55.3 +/- 4.9%, n = 10; p = 0.002). Accumulation of neutrophils within the ischemic region, as assessed by myeloperoxidase activity, declined by 45% (p < 0.05) in AAR from NHEP-treated animals compared to AAR from vehicle-treated animals. Levels of MAC and IL-8 obtained from AAR were less in NHEP-pretreated animals compared to controls. These results suggest that NHEP may protect the myocardium by inhibiting complement activation and subsequent neutrophil infiltration.
Sigma-Aldrich
N-Acetylheparin sodium salt, ~90%