Merck
Nucleosides of azathioprine and thiamiprine as antiarthritics.
Journal of medicinal chemistry (1989-07-01)
T A Krenitsky, W W Hall, J L Selph, J F Truax, R Vinegar
ABSTRACT
Azathioprine [Imuran; 6-[(1-methyl-4-nitro-1H-imidazol-5-yl)thio]-1H-purine] is a widely used immunosuppressive and antiarthritic drug. For the sake of comparison, the riboside, the 2'-deoxyriboside, and the arabinoside of azathioprine and its 2-amino congener, thiamiprine, were prepared by an enzymatic method. In vitro, the cytotoxicities of these aglycons and their nucleosides were similar (ED50 = 0.8-2 microM), except for the arabinosides, which were nontoxic (ED50 greater than 100 microM). In vivo, their activities were compared in the rat adjuvant arthritis model. The ribosides and 2'-deoxyribosides were less potent than their corresponding aglycons. The safety indexes of these nucleosides were comparable to those of the corresponding aglycons except for the 2'-deoxyriboside of azathioprine, which had an appreciably lower safety index than did azathioprine. Both arabinosides were inactive and nontoxic. All of the aglycons tested (6-mercaptopurine, azathioprine, 6-thioguanine, and thiamiprine) were of similar potency. However, azathioprine had a more favorable therapeutic index than did 6-mercaptopurine. Similarly, thiamiprine was safer than was 6-thioguanine. In this model, the S-(1-methyl-4-nitro-1H-imidazol-5-yl) moiety imparted greater safety to these thiopurines by decreasing toxicity but not affecting potency.
Product Description
Azathioprine impurity G, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.