Merck
  • Development of Antibacterial Ti-Cu x Alloys for Dental Applications: Effects of Ageing for Alloys with Up to 10 wt% Cu.
Development of Antibacterial Ti-Cu x Alloys for Dental Applications: Effects of Ageing for Alloys with Up to 10 wt% Cu.
Materials (Basel, Switzerland) (2019-12-11)
Lee Fowler, Nomsombuluko Masia, Lesley A Cornish, Lesley H Chown, Håkan Engqvist, Susanne Norgren, Caroline Öhman-Mägi
ABSTRACT
Peri-implantitis, a disease caused by bacteria, affects dental implants in patients. It is widely treated with antibiotics, however, with growing antibiotic resistance new strategies are required. Titanium-copper alloys are prospective antibacterial biomaterials, with the potential to be a remedy against peri-implantitis and antibiotic resistance. The aim of this study was to investigate Ti-Cux alloys, exploring how Cu content (up to 10 wt%) and ageing affect the material properties. Electron microscopy, X-ray diffraction, hardness testing, bacteriological culture, and electrochemical testing were employed to characterize the materials. It was found that alloys with above 3 wt% Cu had two phases and ageing increased the volume fraction of Ti2Cu. An un-aged alloy of 5 wt% Cu showed what could be Ti3Cu, in addition to the α-Ti phase. The hardness gradually increased with increased Cu additions, while ageing only affected the alloy with 10 wt% Cu (due to changes in microstructure). Ageing resulted in faster passivation of the alloys. After two hours the aged 10 wt% Cu alloy was the only material with an antibacterial effect, while after six hours, bacteria killing occurred in all alloys with above 5 wt% Cu. In conclusion, it was possible to tune the material and antibacterial properties of Ti-Cux alloys by changing the Cu concentration and ageing, which makes further optimization towards an antibacterial material promising.
Sigma-Aldrich
Copper, rod, diam. 11 mm, 99.9999% trace metals basis