Merck
  • TRAIL receptor gene editing unveils TRAIL-R1 as a master player of apoptosis induced by TRAIL and ER stress.
TRAIL receptor gene editing unveils TRAIL-R1 as a master player of apoptosis induced by TRAIL and ER stress.
Oncotarget (2017-01-01)
Florent Dufour, Thibault Rattier, Andrei Alexandru Constantinescu, Luciana Zischler, Aymeric Morlé, Hazem Ben Mabrouk, Etienne Humblin, Guillaume Jacquemin, Eva Szegezdi, Fabien Delacote, Naziha Marrakchi, Gilles Guichard, Catherine Pellat-Deceunynck, Pierre Vacher, Patrick Legembre, Carmen Garrido, Olivier Micheau
ABSTRACT
TRAIL induces selective tumor cell death through TRAIL-R1 and TRAIL-R2. Despite the fact that these receptors share high structural homologies, induction of apoptosis upon ER stress, cell autonomous motility and invasion have solely been described to occur through TRAIL-R2. Using the TALEN gene-editing approach, we show that TRAIL-R1 can also induce apoptosis during unresolved unfolded protein response (UPR). Likewise, TRAIL-R1 was found to co-immunoprecipitate with FADD and caspase-8 during ER stress. Its deficiency conferred resistance to apoptosis induced by thaspigargin, tunicamycin or brefeldin A. Our data also demonstrate that tumor cell motility and invasion-induced by TRAIL-R2 is not cell autonomous but induced in a TRAIL-dependant manner. TRAIL-R1, on the other hand, is unable to trigger cell migration owing to its inability to induce an increase in calcium flux. Importantly, all the isogenic cell lines generated in this study revealed that apoptosis induced TRAIL is preferentially induced by TRAIL-R1. Taken together, our results provide novel insights into the physiological functions of TRAIL-R1 and TRAIL-R2 and suggest that targeting TRAIL-R1 for anticancer therapy is likely to be more appropriate owing to its lack of pro-motile signaling capability.
Sigma-Aldrich
Anti-DR4 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-DR5 Antibody, CT, Chemicon®, from rabbit