Merck
  • Vault packaged enzyme mediated degradation of amino-aromatic energetic compounds.
Vault packaged enzyme mediated degradation of amino-aromatic energetic compounds.
Chemosphere (2019-10-28)
Anjali G Lothe, Shashank Singh Kalra, Meng Wang, Elizabeth Erin Mack, Claudia Walecka-Hutchison, Valerie A Kickhoefer, Leonard H Rome, Shaily Mahendra
ABSTRACT
Amino-aromatic compounds, 2-amino-4-nitrotoluene (ANT), and 2,4-diaminotoluene (DAT) are carcinogens and environmentally persistent pollutants. In this study, we investigated their degradation by natural manganese peroxidase (nMnP) derived from Phanerochaete chrysosporium and recombinant manganese peroxidase packaged in vaults (vMnP). Encapsulation of manganese peroxidase (MnP) in ribonucleoprotein nanoparticle cages, called vaults, was achieved by creating recombinant vaults in yeast Pichia pastoris. Vault packaging increased the stability of MnP by locally sequestering multiple copies of the enzyme. Within 96  h, both vMnP and nMnP catalyzed over 72% removal of ANT in-vitro, which indicates that vault packaging did not limit substrate diffusion. It was observed that vMnP was more efficient than nMnP and P. chrysosporium for the catalysis of target contaminants. Only 57% of ANT was degraded by P. chrysosporium even when MnP activity reached about 480 U L-1 in cultures. At 1.5 U L-1 initial activity, vMnP achieved 38% of ANT and 51% of DAT degradation, whereas even 2.7 times higher activity of nMnP showed insignificant biodegradation of both compounds. These results imply that due to protection by vault cages, vMnP has lower inactivation rates. Thus, it works effectively at lower dosage for a longer duration compared to nMnP without requiring frequent replenishment. Collectively, these results indicate that fungal enzymes packaged in vault nanoparticles are more stable and active, and they would be effective in biodegradation of energetic compounds in industrial processes, waste treatment, and contaminated environments.
Sigma-Aldrich
(4-Maleimidobutyramidomethyl)polystyrene, extent of labeling: ~0.4 mmol/g "maleimide" loading
Sigma-Aldrich
2-Methyl-5-nitroaniline, 98%