Merck
  • Optical sensor for the detection of caspase-9 activity in a single cell.
Optical sensor for the detection of caspase-9 activity in a single cell.
Journal of the American Chemical Society (2004-03-05)
Paul M Kasili, Joon Myong Song, Tuan Vo-Dinh
ABSTRACT
We demonstrate for the first time, the application and utility of a unique optical sensor having a nanoprobe for monitoring the onset of the mitochondrial pathway of apoptosis in a single living cell by detecting enzymatic activities of caspase-9. Minimally invasive analysis of single live MCF-7 cells for caspase-9 activity is demonstrated using the optical sensor which employs a modification of an immunochemical assay format for the immobilization of nonfluorescent enzyme substrate, Leucine-GlutamicAcid-Histidine-AsparticAcid-7-amino-4-methylcoumarin (LEHD-AMC). LEHD-AMC covalently attached on the nanoprobe tip of an optical sensor is cleaved during apoptosis by caspase-9 generating free AMC. An evanescent field is used to excite cleaved AMC and the resulting fluorescence signal is detected. By quantitatively monitoring the changes in fluorescence signals, caspase-9 activity within a single living MCF-7 cell was detected. By comparing of the fluorescence signals from apoptotic cells induced by photodynamic treatment and nonapoptotic cells, we successfully detected caspase-9 activity, which indicates the onset of apoptosis in the cells.
Sigma-Aldrich
Caspase Substrate