Merck
  • Effect of Low-Level Laser on Some Metals Related to Redox State and Histological Alterations in the Liver and Kidney of Irradiated Rats.
Effect of Low-Level Laser on Some Metals Related to Redox State and Histological Alterations in the Liver and Kidney of Irradiated Rats.
Biological trace element research (2019-07-18)
Nadia Abdel-Magied, Ahmed A Elkady, Salma M Abdel Fattah
ABSTRACT
Low-level laser therapy (LLLT) is a type of medicine that uses laser light at low levels to activate the cellular chromophores and the initiation of cellular signaling. This study aimed to evaluate the photomodulation effect of LLL against ionizing radiation (IR)-induced metal disorders related to redox state in the liver and kidney of male rats. Rats were divided into 4 groups (control, LLLT, IR (7Gy), IR+LLLT). The results showed that LLLT 870 nm one time for 3 days post-irradiation revealed redistribution of iron (Fe), copper (Cu), zinc (Zn),calcium (Ca), magnesium (Mg), manganese (Mn), and selenium (Se) in the liver and kidney tissues. Moreover, LLLT attenuated the oxidative stress manifested by a marked reduction of hydrogen peroxide (H2O2), 4-hydroxynonenal (4-HNE), total oxidant state (TOS), and oxidative stress index (OSI) associated with a significant increase in total antioxidant status (TAS), glutathione (GSH) content, and glutathione peroxide (GPx), glutathione reductase (GRx), superoxide dismutase(SOD), and catalase (CAT) activities. Moreover, LLLT displayed an increase in glutathione-S-transferase (GSH-T) and ceruloplasmin activities and a decrease in the activity of gamma-glutamyl transferase (γ-GT). Besides, LLLT significantly attenuated the histological changes in the liver and kidney tissues, denoted by a reduction in the necrotic and degenerative changes of hepatocytes and an improvement in the corpuscles and tubules of the kidney. In conclusion, LLLT could be used as an adjuvant treatment post-exposure to radiation, while it is not beneficial to use it on the normal tissue.
Sigma-Aldrich
Ceruloplasmin Activity Colorimetric Kit, Sufficient for 100 Colorimetric tests