Merck
  • Effects of Synergistic Inhibition on α-glucosidase by Phytoalexins in Soybeans.
Effects of Synergistic Inhibition on α-glucosidase by Phytoalexins in Soybeans.
Biomolecules (2019-12-11)
Hyeong-U Son, Eun-Kyeong Yoon, Chi-Yeol Yoo, Chul-Hong Park, Myung-Ae Bae, Tae-Ho Kim, Chang Hyung Lee, Ki Won Lee, Hogyun Seo, Kyung-Jin Kim, Sang-Han Lee
ABSTRACT
To determine the mechanism of action of the effects of phytoalexins in soybeans, we analyzed α-glucosidase inhibition kinetics using Michaelis-Menten plots and Lineweaver-Burk plots. The results showed that the type of inhibition with glyceollin was competitive, that of genistein was noncompetitive, that of daidzein was uncompetitive, and luteolin showed a mixed mode of action. The Ki values were determined using a Dixon plot as glyceollin, 18.99 μM; genistein, 15.42 μM; luteolin, 16.81 μM; and daidzein, 9.99 μM. Furthermore, potential synergistic effects between glyceollin and the three polyphenols were investigated. A combination of glyceollin and luteolin at a ratio of 3:7 exhibited synergistic effects on α-glucosidase inhibition, having a combination index (CI) of 0.64244, according to the CI-isobologram equation. Collectively, these results showed that a combination of glyceollin and luteolin has the potential to inhibit α-glucosidase activity via a synergistic mode of inhibition.
Sigma-Aldrich
4-Nitrophenyl α-D-glucopyranoside, ≥99%