Merck
  • Plant-Mediated Enantioselective Transformation of Indan-1-one and Indan-1-ol. Part 2.
Plant-Mediated Enantioselective Transformation of Indan-1-one and Indan-1-ol. Part 2.
Molecules (Basel, Switzerland) (2019-12-01)
Wanda Mączka, Katarzyna Wińska, Małgorzata Grabarczyk, Renata Galek
ABSTRACT
The main purpose of this publication was to obtain the S-enantiomer of indan-1-ol with high enantiomeric excess and satisfactory yield. In our research, we used carrot callus cultures (Daucuscarota L.), whereby the enzymatic system reduced indan-1-one and oxidized indan-1-ol. During the reaction of reduction, after five days, we received over 50% conversion, with the enantiomeric excess of the formed S-alcohol above 99%. In turn, during the oxidation of racemic indan-1-ol after 15 days, 36.7% of alcohol with an enantiomeric excess 57.4% S(+) remained in the reaction mixture. In addition, our research confirmed that the reactions of reduction and oxidation are competing reactions during the transformation of indan-1-ol and indan-1-one in carrot callus cultures.
Sigma-Aldrich
1-Indanol, 98%
Pricing and availability is not currently available.