Merck
  • Single injection of IL-12 coacervate as an effective therapy against B16-F10 melanoma in mice.
Single injection of IL-12 coacervate as an effective therapy against B16-F10 melanoma in mice.
Journal of controlled release : official journal of the Controlled Release Society (2019-12-24)
Mintai P Hwang, Ronald J Fecek, Tianyue Qin, Walter J Storkus, Yadong Wang
ABSTRACT
Melanoma is the deadliest type of skin cancer with one of the fastest increasing incidence rates among solid tumors. The use of checkpoint inhibitors (e.g. αPD-1 antibody) has recently emerged as a viable alternative to conventional modes of therapy. However, increasing evidence points towards the need for a tumor priming step to improve intratumoral immune cell infiltration. IL-12 is an immune-activating cytokine with such potential and was explored in earlier clinical trials as a highly concentrated systemic infusion. This unfortunately led to severe adverse effects. From this perspective, the localization and gradual release of such a potent immunotherapeutic agent in the tumor microenvironment is desired. This manuscript reports the use of a heparin-based complex coacervate to deliver IL-12, in which heparin-binding motifs on IL-12 allow for its effective encapsulation. IL-12-encapsulated complex coacervates significantly improved the bioactivity of IL-12 and provided protection from proteolytic cleavage in-vitro. Indeed, a single injection of IL-12 coacervate significantly inhibits the in-vivo growth of treated and untreated, contralateral tumor growth in a syngeneic B16F10 mouse melanoma model. Furthermore, tumors in mice receiving IL-12 complex coacervate treatment displayed increased infiltration by natural killer (NK) cells and CD8α+ T cells, and a decreased presence of CD4+Foxp3+ regulatory T cells. This study provides proof-of-concept data supporting the use of complex coacervates for sustained delivery of immunostimulatory proteins as an effective therapeutic strategy against disseminated tumors.
Sigma-Aldrich
Boc-Arg-OH
Sigma-Aldrich
Boc-Asp-OH, 99%