Merck
  • TNFα-induced Up-regulation of Ascl2 Affects the Differentiation and Proliferation of Neural Stem Cells.
TNFα-induced Up-regulation of Ascl2 Affects the Differentiation and Proliferation of Neural Stem Cells.
Aging and disease (2019-12-04)
Zhongfeng Liu, Xuan Wang, Kewen Jiang, Xunming Ji, Y Alex Zhang, Zhiguo Chen
ABSTRACT
The molecular mediators underlying the effects of inflammation on neural stem cells (NSCs) are not fully characterized. In this study, we identified Ascl2 as a downstream basic helix-loop-helix (bHLH) transcription factor in NSCs following exposure to TNFα. Under normal conditions, Ascl2 expression is inhibited at post-transcriptional levels by miR-26a, which targets the 3' untranslated region (UTR) of Ascl2. Upon exposure to TNFα, miR-26a expression is reduced, which leads to up-regulation of Ascl2. Overexpression of Ascl2 promotes neuronal differentiation, reduces proliferation, and increases the level of cleaved CASPASE 3 in NSCs, as observed in the in vitro and in ovo experiments. Ascl2 may serve in NSCs as a standby factor that readily responds to TNFα, which is often induced in inflammatory situations. In a chronic inflammatory condition with consistent up-regulation of TNFα, overexpression of Ascl2 may inhibit neurogenesis as a net result.
Sigma-Aldrich
Anti-Achaete Scute homolog 2 Antibody, clone 7E2, clone 7E2, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Pricing and availability is not currently available.