Merck
  • Folliculin regulates mTORC1/2 and WNT pathways in early human pluripotency.
Folliculin regulates mTORC1/2 and WNT pathways in early human pluripotency.
Nature communications (2019-02-09)
J Mathieu, D Detraux, D Kuppers, Y Wang, C Cavanaugh, S Sidhu, S Levy, A M Robitaille, A Ferreccio, T Bottorff, A McAlister, L Somasundaram, F Artoni, S Battle, R D Hawkins, R T Moon, C B Ware, P J Paddison, H Ruohola-Baker
ABSTRACT
To reveal how cells exit human pluripotency, we designed a CRISPR-Cas9 screen exploiting the metabolic and epigenetic differences between naïve and primed pluripotent cells. We identify the tumor suppressor, Folliculin(FLCN) as a critical gene required for the exit from human pluripotency. Here we show that FLCN Knock-out (KO) hESCs maintain the naïve pluripotent state but cannot exit the state since the critical transcription factor TFE3 remains active in the nucleus. TFE3 targets up-regulated in FLCN KO exit assay are members of Wnt pathway and ESRRB. Treatment of FLCN KO hESC with a Wnt inhibitor, but not ESRRB/FLCN double mutant, rescues the cells, allowing the exit from the naïve state. Using co-immunoprecipitation and mass spectrometry analysis we identify unique FLCN binding partners. The interactions of FLCN with components of the mTOR pathway (mTORC1 and mTORC2) reveal a mechanism of FLCN function during exit from naïve pluripotency.
Sigma-Aldrich
Anti-TFE3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Sin1 Antibody, clone 1C7.2, clone 1C7.2, from mouse