Merck
  • Novel targets for endoplasmic reticulum stress-induced apoptosis in B-CLL.
Novel targets for endoplasmic reticulum stress-induced apoptosis in B-CLL.
Blood (2010-07-16)
Emanuela Rosati, Rita Sabatini, Giuliana Rampino, Filomena De Falco, Mauro Di Ianni, Franca Falzetti, Katia Fettucciari, Andrea Bartoli, Isabella Screpanti, Pierfrancesco Marconi
ABSTRACT
A better understanding of apoptotic signaling in B-chronic lymphocytic leukemia (B-CLL) cells may help to define new therapeutic strategies. This study investigated endoplasmic reticulum (ER) stress signaling in spontaneous apoptosis of B-CLL cells and whether manipulating ER stress increases their apoptosis. Results show that a novel ER stress-triggered caspase cascade, initiated by caspase-4 and involving caspase-8 and -3, plays an important role in spontaneous B-CLL cell apoptosis. ER stress-induced apoptosis in B-CLL cells also involves CHOP/GADD153 up-regulation, increased JNK1/2 phosphorylation, and caspase-8-mediated cleavage of Bap31 to Bap20, known to propagate apoptotic signals from ER to mitochondria. In ex vivo B-CLL cells, some apoptotic events associated with mitochondrial pathway also occur, including mitochondrial cytochrome c release and caspase-9 processing. However, pharmacologic inhibition studies show that caspase-9 plays a minor role in B-CLL cell apoptosis. ER stress also triggers survival signals in B-CLL cells by increasing BiP/GRP78 expression. Manipulating ER signaling by siRNA down-regulation of BiP/GRP78 or treating B-CLL cells with 2 well-known ER stress-inducers, tunicamycin and thapsigargin, increases their apoptosis. Overall, our findings show that ER triggers an essential pathway for B-CLL cell apoptosis and suggest that genetic and pharmacologic manipulation of ER signaling could represent an important therapeutic strategy.
Sigma-Aldrich
Anti-Caspase 9 Antibody, clone 96-2-22, clone 96-2-22, Upstate®, from mouse