Merck
  • MicroRNA‑20a promotes inflammation via the nuclear factor‑κB signaling pathway in pediatric pneumonia.
MicroRNA‑20a promotes inflammation via the nuclear factor‑κB signaling pathway in pediatric pneumonia.
Molecular medicine reports (2017-11-09)
Zhihong Liu, Haiying Yu, Qiuye Guo
ABSTRACT
Pneumonia is a common respiratory disease worldwide, which is preventable and treatable; however, it is recognized as a leading cause of mortality in children. The present study aimed to investigate the role and mechanism of microRNA (miR)‑20a in inflammation in pediatric pneumonia. Clinical serum samples were collected from children with pneumonia and healthy children. Initially, the serum expression levels of miR‑20a were detected by reverse transcription‑quantitative polymerase chain reaction. Subsequently, A549 cells were randomly divided into four groups: Control group; lipopolysaccharide (LPS; 1 µg/ml) group; LPS + miR‑20a group; and LPS + miR‑20a + pyrrolidine dithiocarbamate (PDTC; 100 mmol/l) group. The concentrations of interleukin‑6 (IL‑6), tumor necrosis factor (TNF)‑α and C‑reactive protein (CRP) in clinical serum samples and A549 cells were determined by ELISA. In addition, the protein expression levels of inhibitor of nuclear factor (NF)‑κB α (IκBα) and phosphorylated (p)‑NF‑κB were measured by western blotting. The results demonstrated that miR‑20a was upregulated in children with pneumonia and in lung cells with LPS‑induced inflammatory injury (P<0.01). In addition, compared with the LPS group, cells in the LPS + miR‑20a group exhibited increased expression levels of IL‑6, TNF‑α and CRP (P<0.05). Overexpression of miR‑20a also resulted in upregulation of the expression levels of IκBα and p‑NF‑κB compared with in the LPS group (P<0.05). Furthermore, treatment with the NF‑κB inhibitor PDTC inhibited the expression of inflammatory factors compared with in the LPS + miR‑20a group (P<0.05). In conclusion, the present study indicated that miR‑20a is upregulated in pediatric pneumonia, and overexpression of miR‑20a may promote inflammation through activation of the NF‑κB signaling pathway.
Sigma-Aldrich
Anti-phospho-NF-κB p65 (pSer281) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-IκB-α antibody produced in rabbit, affinity isolated antibody