Merck
  • Single-molecule sensing of peptides and nucleic acids by engineered aerolysin nanopores.
Single-molecule sensing of peptides and nucleic acids by engineered aerolysin nanopores.
Nature communications (2019-10-31)
Chan Cao, Nuria Cirauqui, Maria Jose Marcaida, Elena Buglakova, Alice Duperrex, Aleksandra Radenovic, Matteo Dal Peraro
ABSTRACT
Nanopore sensing is a powerful single-molecule approach for the detection of biomolecules. Recent studies have demonstrated that aerolysin is a promising candidate to improve the accuracy of DNA sequencing and to develop novel single-molecule proteomic strategies. However, the structure-function relationship between the aerolysin nanopore and its molecular sensing properties remains insufficiently explored. Herein, a set of mutated pores were rationally designed and evaluated in silico by molecular simulations and in vitro by single-channel recording and molecular translocation experiments to study the pore structural variation, ion selectivity, ionic conductance and capabilities for sensing several biomolecules. Our results show that the ion selectivity and sensing ability of aerolysin are mostly controlled by electrostatics and the narrow diameter of the double β-barrel cap. By engineering single-site mutants, a more accurate molecular detection of nucleic acids and peptides has been achieved. These findings open avenues for developing aerolysin nanopores into powerful sensing devices.
Product Description
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand