Merck
  • Facile preparation of biomass lignin-based hydroxyethyl cellulose super-absorbent hydrogel for dye pollutant removal.
Facile preparation of biomass lignin-based hydroxyethyl cellulose super-absorbent hydrogel for dye pollutant removal.
International journal of biological macromolecules (2019-07-08)
Siqi Huang, Linjun Wu, Tianzhang Li, Danyuan Xu, Xuliang Lin, Chuande Wu
ABSTRACT
The severe preparation process, poor swelling properties and mechanical properties of traditional cellulose and polyvinyl alcohol (PVA) composite hydrogels heavily limited their practical applications. To solve these issues, we use long-chain hydroxyethyl celluloses (HECs) as framework backbones, short-chain PVAs as branched chains, lignin molecules as extended crosslinkers and epichlorohydrin molecules as crosslinkers to prepare the lignin-based hydroxyethyl cellulose-PVA (LCP) super-absorbent hydrogels in the alkaline aqueous solution under mild reaction conditions, demonstrating high swelling ratio of up to 1220 g/g. The LCP hydrogels could take up large amounts of positively charged dyes rhodamine 6G, crystal violet and methylene blue with uptakes of 153, 184 and 196 mg/g, respectively. The LCP super-absorbent hydrogels also present excellent water retention, biodegradability and excellent swelling properties, which are very promising for applications in the fields of commercial diapers, soil water retention and seed cultivation in agriculture, and dye pollutant removal.
Sigma-Aldrich
1-(p-Tolyl)ethanol, ≥97.0% (GC)