Merck
  • Neural anti-inflammatory action mediated by two types of acetylcholine receptors in the small intestine.
Neural anti-inflammatory action mediated by two types of acetylcholine receptors in the small intestine.
Scientific reports (2019-04-12)
Hitomi Kimura, Yu-Ki Imura, Hirotaka Tomiyasu, Taiki Mihara, Noriyuki Kaji, Koichi Ohno, Toshihiro Unno, Yasuyuki Tanahashi, Tong-Rong Jan, Hirokazu Tsubone, Hiroshi Ozaki, Masatoshi Hori
ABSTRACT
Gastrointestinal prokinetic agents function as serotonin-4 receptor (5-HT4R) agonists to activate myenteric plexus neurons to release acetylcholine (ACh), which then induce anti-inflammatory action. Details of this pathway, however, remain unknown. The aim of this study is to clarify the anti-inflammatory mechanism underlying the 5-HT4R agonist, mosapride citrate (MOS)-induced anti-inflammatory action on postoperative ileus (POI). POI models were generated from wild-type C57BL6/J (WT), 5-HT4R knock-out (S4R KO), α7 nicotinic AChR KO (α7 R KO), and M2 muscarinic ACh receptor KO (M2R KO) mice. MOS attenuated leukocyte infiltration in WT. MOS-induced anti-inflammatory action was completely abolished in both S4R KO and S4R KO mice upon wild-type bone marrow transplantation. MOS-induced anti-inflammatory action against macrophage infiltration, but not neutrophil infiltration, was attenuated in α7 R KO mice. Selective α7nAChR agonists (PNU-282987 and AR-R17779) also inhibited only macrophage infiltration in POI. MOS-mediated inhibition of neutrophil infiltration was diminished by atropine, M2AChR antagonist, methoctramine, and in M2R KO mice. Stimulation with 5-HT4R inhibits leukocyte infiltration in POI, possibly through myenteric plexus activation. Released ACh inhibited macrophage and neutrophil infiltration likely by activation of α7nAChR on macrophages and M2AChR. Thus, macrophage and neutrophil recruitment into inflamed sites is regulated by different types of AChR in the small intestine.