- Nickel-Catalyzed Decarbonylative Stannylation of Acyl Fluorides under Ligand-Free Conditions.
Nickel-Catalyzed Decarbonylative Stannylation of Acyl Fluorides under Ligand-Free Conditions.
Molecules (Basel, Switzerland) (2019-05-01)
Xiu Wang, Zhenhua Wang, Li Liu, Yuya Asanuma, Yasushi Nishihara
PMID31035405
ABSTRACT
Nickel-catalyzed decarbonylative stannylation of acyl fluorides under ligand-free conditions was disclosed. A variety of aromatic acyl fluorides are capable of reacting with silylstannanes in the presence of cesium fluoride. A one-pot decarbonylative stannylation/Migita-Kosugi-Stille reaction of benzoyl fluoride, giving rise to the direct formation of the corresponding cross-coupled products, further demonstrated the synthetic utility of the present method. This newly developed methodology with a good functional-group compatibility via C-F bond cleavage and C-Sn bond formation under nickel catalysis opens a new area for the functionalization of acyl fluorides in terms of carbon-heteroatom bond formation.