Merck
Nickel-catalyzed Suzuki-Miyaura reaction of aryl fluorides.
Journal of the American Chemical Society (2011-10-26)
Mamoru Tobisu, Tian Xu, Toshiaki Shimasaki, Naoto Chatani
ABSTRACT
Two protocols for the nickel-catalyzed cross-coupling of aryl fluorides with aryl boronic esters have been developed. The first employs metal fluoride cocatalysts, such as ZrF(4) and TiF(4), which enable Suzuki-Miyaura reactions of aryl fluorides bearing electron-withdrawing (ketones, esters, and CF(3)), aryl and alkenyl groups as well as those comprising fused aromatic rings, such as fluoronaphthalenes and fluoroquinolines. The second protocol employs aryl fluorides bearing ortho-directing groups, which facilitate the difficult C-F bond activation process via cyclometalation. N-heterocycles, such as pyridines, quinolines, pyrazoles, and oxazolines, can successfully promote cross-coupling with an array of organoboronic esters. A study into the substituent effects with respect to both coupling components has provided fundamental insights into the mechanism of the nickel-catalyzed cross-coupling of aryl fluorides.
Sigma-Aldrich
4-(Dimethylamino)phenylboronic acid, ≥95.0%