Merck
  • New organic dye based on a 3,6-disubstituted carbazole donor for efficient dye-sensitized solar cells.
New organic dye based on a 3,6-disubstituted carbazole donor for efficient dye-sensitized solar cells.
Chemistry, an Asian journal (2011-12-14)
Woochul Lee, Nara Cho, Jongchul Kwon, Jaejung Ko, Jong-In Hong
ABSTRACT
We have synthesized and characterized four organic dyes (H1-H4) based on a 3,6-disubstituted carbazole donor as sensitizers in dye-sensitized solar cells. These dyes have high molar extinction coefficients and energy levels suitable for electron transfer from an electrolyte to nanocrystalline TiO(2) particles. Under standard air mass 1.5 global (AM 1.5 G) solar irradiation, a device using dye H4 exhibits a short-circuit current density (J(sc)) of 13.7 mA cm(-2), an open-circuit voltage (V(oc)) of 0.68 V, a fill factor (FF) of 0.70, and a calculated efficiency of 6.52%. This performance is comparable to that of a reference cell based on N719 (7.30%) under the same conditions. After 1000 hours of visible-light soaking at 60 °C, the overall efficiency remained at 95% of the initial value.
Sigma-Aldrich
4-(Diphenylamino)phenylboronic acid, ≥95%
Pricing and availability is not currently available.