Merck
Inhibition of cytochromes P450 by antifungal imidazole derivatives.
Drug metabolism and disposition: the biological fate of chemicals (2002-02-21)
Wenjiang Zhang, Yamini Ramamoorthy, Tansel Kilicarslan, Helma Nolte, Rachel F Tyndale, Edward M Sellers
ABSTRACT
The interactions of a panel of antifungal agents with cytochromes P450 (P450s), as a means of predicting potential drug-drug interactions, have not yet been investigated. The objective of this study was to evaluate the specificity and selectivity of five antifungal agents using selective probe reactions for each of the eight major P450s. The index reactions used were phenacetin O-deethylation (for CYP1A2), coumarin 7-hydroxylation (CYP2A6), diclofenac 4'-hydroxylation (CYP2C9), omeprazole 5-hydroxylation (CYP2C19), dextromethorphan O-demethylation (CYP2D6), 7-ethoxy-4-trifluoromethylcoumarin deethylation (CYP2B6), chlorzoxazone 6-hydroxylation (CYP2E1), and omeprazole sulfonation (CYP3A4). Five antifungal agents that include an imidazole moiety (clotrimazole, miconazole, sulconazole, tioconazole, and ketoconazole) were examined in cDNA-expressing microsomes from human lymphoblast cells or human liver microsomes. All inhibitors studied demonstrated nonselective inhibition of P450s. Ketoconazole seemed to be the most selective for CYP3A4, although it also inhibited CYP2C9. High-affinity inhibition was seen for CYP1A2 (sulconazole and tioconazole K(i), 0.4 microM), CYP2B6 (miconazole K(i), 0.05 microM; sulconazole K(i), 0.04 microM), CYP2C19 (miconazole K(i), 0.05 microM; sulconazole K(i), 0.008 microM; tioconazole K(i), 0.04 microM), CYP2C9 (sulconazole K(i), 0.01 microM), CYP2D6 (miconazole K(i), 0.70 microM; sulconazole K(i), 0.40 microM), CYP2E1 (tioconazole K(i), 0.4 microM), and CYP3A4 (clotrimazole K(i), 0.02 microM; miconazole K(i), 0.03 microM; tioconazole K(i), 0.02 microM). Therefore, this class of compounds is likely to result in significant drug-drug interactions in vivo.
Sigma-Aldrich
Tioconazole, 97%
Pricing and availability is not currently available.