Merck
  • Overexpression of the lung cancer-prognostic miR-146b microRNAs has a minimal and negative effect on the malignant phenotype of A549 lung cancer cells.
Overexpression of the lung cancer-prognostic miR-146b microRNAs has a minimal and negative effect on the malignant phenotype of A549 lung cancer cells.
PloS one (2011-07-27)
Santosh Kumar Patnaik, Eric Kannisto, Reema Mallick, Sai Yendamuri
ABSTRACT
Expression levels of miR-146b-5p and -3p microRNAs in human non-small cell lung cancer (NSCLC) are associated with recurrence of the disease after surgery. To understand this, the effect of miR-146b overexpression was studied in A549 human lung cancer cells. A549 cells, engineered with lentiviruses to overexpress the human pre-miR-146b precursor microRNA, were examined for proliferation, colony formation on plastic surface and in soft agar, migration and invasiveness in cell culture and in vivo in mice, chemosensitivity to cisplatin and doxorubicin, and global gene expression. miR-146b expressions were assessed in microdissected stroma and epithelia of human NSCLC tumors. Association of miR-146b-5p and -3p expression in early stage NSCLC with recurrence was analyzed. A549 pre-miR-146b-overexpressors had 3-8-fold higher levels of both miR-146b microRNAs than control cells. Overexpression did not alter cellular proliferation, chemosensitivity, migration, or invasiveness; affected only 0.3% of the mRNA transcriptome; and, reduced the ability to form colonies in vitro by 25%. In human NSCLC tumors, expression of both miR-146b microRNAs was 7-10-fold higher in stroma than in cancerous epithelia, and higher miR-146b-5p but lower -3p levels were predictive of recurrence. Only a minimal effect of pre-miR-146b overexpression on the malignant phenotype was seen in A549 cells. This could be because of opposing effects of miR-146b-5p and -3p overexpression as suggested by the conflicting recurrence-predictive values of the two microRNAs, or because miR-146b expression changes in non-cancerous stroma and not cancerous epithelia of tumors are responsible for the prognostic value of miR-146b.
Product Description
Millipore
Scepter 2.0 Handheld Automated Cell Counter, Rapid cell counts in a handheld easy to use format, includes pkg of 40 μm Scepter Cell Counter Sensors