Merck
  • Kleisins: a superfamily of bacterial and eukaryotic SMC protein partners.
Kleisins: a superfamily of bacterial and eukaryotic SMC protein partners.
Molecular cell (2003-04-02)
Alexander Schleiffer, Susanne Kaitna, Sebastian Maurer-Stroh, Michael Glotzer, Kim Nasmyth, Frank Eisenhaber
ABSTRACT
We describe a superfamily of eukaryotic and prokaryotic proteins (kleisins) that includes ScpA, Scc1, Rec8, and Barren. Scc1 interacts with SMC proteins through N- and C-terminal domains to form a ring-like structure. Since these are the only domains conserved among kleisins, we suggest that ring formation with SMC proteins may define this family.