Merck
  • Therapeutic potential of neuronal nicotinic acetylcholine receptor agonists as novel analgesics.
Therapeutic potential of neuronal nicotinic acetylcholine receptor agonists as novel analgesics.
Biochemical pharmacology (1999-10-06)
M W Decker, M D Meyer
ABSTRACT
Pharmacological treatments for pain have come largely from two classes of compounds--the opioids and the nonsteroidal anti-inflammatory drugs (NSAIDs). Because of deficiencies associated with these two classes of compounds, exploration of novel approaches to pain relief has intensified of late. Nicotine, a neuronal nicotinic acetylcholine receptor (nAChR) agonist, has long been known to have antinociceptive effects in both experimental animals and humans. The relatively modest antinociceptive effects and the toxicities associated with nicotine preclude its development as an analgesic agent. However, recent discoveries in the nAChR field have stimulated interest in nAChR-targeted compounds as potential analgesic agents. Epibatidine, a potent nAChR agonist, was found to have full efficacy relative to opioids in preclinical pain models. Although epibatidine is toxic, these observations demonstrated that modest efficacy is not a general limitation of nAChR agonists. Moreover, exploration of the molecular biology of nAChRs revealed evidence of receptor diversity, suggesting that nAChR subtype-selective agents less toxic than nicotine might be discovered; and early medicinal chemistry efforts already have resulted in compounds with improved safety profiles. For example, ABT-594 is a nAChR agonist with the antinociceptive efficacy of epibatidine, but with an improved safety profile. This commentary reviews recent findings with nAChR-targeted compounds, explores potential mechanisms responsible for nAChR-mediated antinociception, and raises issues that must be addressed in developing compounds of this class as analgesics.
Sigma-Aldrich
(±)-Epibatidine dihydrochloride hydrate, ≥98% (HPLC), powder