Merck
  • Mass spectral analysis of neuropeptide expression and distribution in the nervous system of the lobster Homarus americanus.
Mass spectral analysis of neuropeptide expression and distribution in the nervous system of the lobster Homarus americanus.
Journal of proteome research (2009-12-23)
Ruibing Chen, Xiaoyue Jiang, Maria C Prieto Conaway, Iman Mohtashemi, Limei Hui, Rosa Viner, Lingjun Li
ABSTRACT
The lobster Homarus americanus has long served as an important animal model for electrophysiological and behavioral studies. Using this model, we performed a comprehensive investigation of the neuropeptide expression and their localization in the nervous system, which provides useful insights for further understanding of their biological functions. Using nanoLC ESI Q-TOF MS/MS and three types of MALDI instruments, we analyzed the neuropeptide complements in a major neuroendocrine structure, pericardial organ. A total of 57 putative neuropeptides were identified and 18 of them were de novo sequenced. Using direct tissue/extract analysis and bioinformatics software SpecPlot, we charted the global distribution of neuropeptides throughout the nervous system in H. americanus. Furthermore, we also mapped the localization of several neuropeptide families in the brain by high mass resolution and high mass accuracy mass spectrometric imaging (MSI) using a MALDI LTQ Orbitrap mass spectrometer. We have also compared the utility and instrument performance of multiple mass spectrometers for neuropeptide analysis in terms of peptidome coverage, sensitivity, mass spectral resolution and capability for de novo sequencing.
Product Description
Supelco
ProteoMass MALDI Calibration Kit for Exactive Series MS, LTQ XL and LTQ Hybrids, MALDI MS Calibration Standard
Pricing and availability is not currently available.