Merck
  • GAPDH, a novel regulator of the pro-apoptotic mitochondrial membrane permeabilization.
GAPDH, a novel regulator of the pro-apoptotic mitochondrial membrane permeabilization.
Oncogene (2006-10-31)
A Tarze, A Deniaud, M Le Bras, E Maillier, D Molle, N Larochette, N Zamzami, G Jan, G Kroemer, C Brenner
ABSTRACT
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a pleiotropic enzyme that is overexpressed in apoptosis and in several human chronic pathologies. Here, we report that the protein accumulates in mitochondria during apoptosis, and induces the pro-apoptotic mitochondrial membrane permeabilization, a decisive event of the intrinsic pathway of apoptosis. GAPDH was localized by immunogold labeling and identified by matrix-assisted laser desorption/ionization-time of flight and nano liquid chromatography mass spectroscopy/mass spectroscopy in the mitochondrion of various tissues and origins. In isolated mitochondria, GAPDH can be imported and interact with the voltage-dependent anion channel (VDAC1), but not the adenine nucleotide translocase (ANT). The protein mediates a cyclosporin A-inhibitable permeability transition, characterized by a loss of the inner transmembrane potential, matrix swelling, permeabilization of the inner mitochondrial membrane and the release of two pro-apoptotic proteins, cytochrome c and apoptosis-inducing factor (AIF). This novel function of GAPDH might have implications for the understanding of mitochondrial biology, oncogenesis and apoptosis.
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), lyophilized powder, 70-140 units/mg protein