Merck
  • High serum Ephrin-B2 levels predict poor prognosis for patients with gastric cancer.
High serum Ephrin-B2 levels predict poor prognosis for patients with gastric cancer.
Oncology letters (2018-09-15)
Wen-Jing Yue, Yi-Pin Liu, Ming Li, Cheng-Xia Liu, Shao-Jia Mou, Qian-Kun Li, Zi-Ping Chen
ABSTRACT
Gastric cancer is an intractable disease with a poor prognosis and limited treatment options. Its treatment remains a major clinical challenge worldwide. Ephrin-B2 is upregulated and involved in tumor growth in various types of cancer. However, the association between ephrin-B2 and prognosis of gastric cancer, and the potential of ephrin-B2 as a therapeutic target remains unknown. The present study investigated ephrin-B2 as a prognostic factor and a therapeutic target for gastric cancer. Reverse transcription-quantitative polymerase chain reaction was performed to detect the protein expression level of ephrin-B2 in gastric cancer serum samples (n=162) and healthy serum samples (n=165). It was revealed that the protein expression level of ephrin-B2 was significantly upregulated in gastric cancer serum samples compared with the healthy samples. Ephrin-B2 protein expression was associated with tumor size (P<0.001), metastasis (P=0.02) and TNM stage (P=0.03), and was indicated to be an independent prognostic factor for gastric cancer. Furthermore, the Kaplan-Meier survival curve demonstrated that patients with high ephrin-B2 protein expression had shorter overall and progression-free survival rates than those with low ephrin-B2 protein expression. Ephrin-B2 protein expression was induced by small interfering RNA (siRNA) transfection of HGC27 and MKN-45 cells, significantly impeding cell viability and inducing apoptosis of HGC27 and MKN-45 cells compared with the respective negative control (NC) group. Thus, to the best of our knowledge, the present study indicates that ephrin-B2 functions as an oncogene in gastric cancer, and that serum ephrin-B2 level may be a promising non-invasive prognostic indicator, as well as a therapeutic target for gastric cancer.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
GenElute Plasma/Serum RNA Purification Midi Kit