Merck
  • Fixation and Spread of Somatic Mutations in Adult Human Colonic Epithelium.
Fixation and Spread of Somatic Mutations in Adult Human Colonic Epithelium.
Cell stem cell (2018-05-22)
Anna M Nicholson, Cora Olpe, Alice Hoyle, Ann-Sofie Thorsen, Teja Rus, Mathilde Colombé, Roxanne Brunton-Sim, Richard Kemp, Kate Marks, Phil Quirke, Shalini Malhotra, Rogier Ten Hoopen, Ashraf Ibrahim, Cecilia Lindskog, Meagan B Myers, Barbara Parsons, Simon Tavaré, Mark Wilkinson, Edward Morrissey, Douglas J Winton
ABSTRACT
We investigated the means and timing by which mutations become fixed in the human colonic epithelium by visualizing somatic clones and mathematical inference. Fixation requires two sequential steps. First, one of approximately seven active stem cells residing within each colonic crypt has to be mutated. Second, the mutated stem cell has to replace neighbors to populate the entire crypt in a process that takes several years. Subsequent clonal expansion due to crypt fission is infrequent for neutral mutations (around 0.7% of all crypts undergo fission in a single year). Pro-oncogenic mutations subvert both stem cell replacement to accelerate fixation and clonal expansion by crypt fission to achieve high mutant allele frequencies with age. The benchmarking of these behaviors allows the advantage associated with different gene-specific mutations to be compared irrespective of the cellular mechanisms by which they are conferred.