Merck
  • Electroacupuncture at LI11 promotes jejunal motility via the parasympathetic pathway.
Electroacupuncture at LI11 promotes jejunal motility via the parasympathetic pathway.
BMC complementary and alternative medicine (2017-06-24)
Xuanming Hu, Mengqian Yuan, Yin Yin, Yidan Wang, Yuqin Li, Na Zhang, Xueyi Sun, Zhi Yu, Bin Xu
ABSTRACT
Gastrointestinal motility disorder has been demonstrated to be regulated by acupuncture treatment. The mechanisms underlying the effects of acupuncture stimulation of abdominal and lower limb acupoints on gastrointestinal motility have been thoroughly studied; however, the physiology underlying the effects of acupuncture on the forelimbs to mediate gastrointestinal motility requires further exploration. The aim of this study was to determine whether electroacupuncture (EA) at LI11 promotes jejunal motility, whether the parasympathetic pathway participates in this effect, and if so, which somatic afferent nerve fibres are involved. A manometric balloon was used to observe jejunal motility. The effects and mechanisms of EA at LI11 were explored in male Sprague-Dawley rats with or without drug administration (propranolol, clenbuterol, acetylcholine, and atropine) and with or without vagotomy. Three types of male mice (β1β2 receptor-knockout [β1β2-/-] mice, M2M3 receptor-knockout [M2M3-/-] mice and wild-type [WT] mice) were also studied by using different EA intensities (1, 2, 4, 6, and 8 mA). A total of 72 rats and 56 mice were included in the study. EA at LI11 increased the contractile amplitude of jejunal motility in the majority of both rats and mice. However, EA at LI11 did not enhance jejunal motility in rats administered atropine, rats that underwent vagotomy, and M2M3-‍‍/- mice (at all intensities). In WT mice, EA at LI11 significantly increased jejunal motility at all intensities except 1 mA, and a plateau was reached at intensities greater than 4 mA. Our results suggest that EA at LI11 promotes jejunal motility primarily by exciting the parasympathetic pathway, and that Aδ-fibres and C-fibres may play important roles in the process.
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Monoclonal Anti-Pinin antibody produced in mouse, ~1.5 mg/mL, clone 5F1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Atropine, ≥99% (TLC), powder