Merck
  • Identification of the molecular target of small molecule inhibitors of HDL receptor SR-BI activity.
Identification of the molecular target of small molecule inhibitors of HDL receptor SR-BI activity.
Biochemistry (2007-12-11)
Thomas J F Nieland, Jared T Shaw, Firoz A Jaipuri, Jay L Duffner, Angela N Koehler, Sotirios Banakos, Vassilis I Zannis, Tomas Kirchhausen, Monty Krieger
ABSTRACT
Scavenger receptor, class B, type I (SR-BI), controls high-density lipoprotein (HDL) metabolism by mediating cellular selective uptake of lipids from HDL without the concomitant degradation of the lipoprotein particle. We previously identified in a high-throughput chemical screen of intact cells five compounds (BLT-1-5) that inhibit SR-BI-dependent lipid transport from HDL, but do not block HDL binding to SR-BI on the cell surface. Although these BLTs are widely used to examine the diverse functions of SR-BI, their direct target(s), SR-BI itself or some other component of the SR-BI pathway, has not been identified. Here we show that SR-BI in the context of a membrane lipid environment is the target of BLT-1, -3, -4, and -5. The analysis using intact cells and an in vitro system of purified SR-BI reconstituted into liposomes was aided by information derived from structure-activity relationship (SAR) analysis of the most potent of these BLTs, the thiosemicarbazone BLT-1. We found that the sulfur atom of BLT-1 was crucially important for its inhibitory activity, because changing it to an oxygen atom resulted in the isostructural, but essentially inactive, semicarbazone derivative BLT-1sc. SAR analysis also established the importance of BLT-1's hydrophobic tail. BLTs and their corresponding inactive compounds can be used to explore the mechanism and function of SR-BI-mediated selective lipid uptake in diverse mammalian experimental models. Consequently, BLTs may help determine the therapeutic potential of SR-BI-targeted pharmaceutical drugs.
Sigma-Aldrich
(R)-(+)-Warfarin, ≥97% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Naproxen sodium, 98.0-102.0%
Pricing and availability is not currently available.
Sigma-Aldrich
HDL Receptor SR-BI Inhibitor, BLT-1, The HDL Receptor SR-BI Inhibitor, BLT-1, also referenced under CAS 321673-30-7, controls the biological activity of HDL Receptor SR-B. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Pricing and availability is not currently available.