Merck
  • Deprotonative metallation of ferrocenes using mixed lithium-zinc and lithium-cadmium combinations.
Deprotonative metallation of ferrocenes using mixed lithium-zinc and lithium-cadmium combinations.
Chemical communications (Cambridge, England) (2010-04-07)
Gandrath Dayaker, Aare Sreeshailam, Floris Chevallier, Thierry Roisnel, Palakodety Radha Krishna, Florence Mongin
ABSTRACT
A mixed lithium-cadmium amide and a combination of lithium and zinc amides were reacted with a range of ferrocenes; deprotonative mono- or dimetallation in general occurred chemoselectively at room temperature, as evidenced by subsequent quenching with iodine.