Merck
  • Dietary lariciresinol attenuates mammary tumor growth and reduces blood vessel density in human MCF-7 breast cancer xenografts and carcinogen-induced mammary tumors in rats.
Dietary lariciresinol attenuates mammary tumor growth and reduces blood vessel density in human MCF-7 breast cancer xenografts and carcinogen-induced mammary tumors in rats.
International journal of cancer (2008-06-06)
Niina M Saarinen, Anni Wärri, Ruud P M Dings, Maarit Airio, Annika I Smeds, Sari Mäkelä
ABSTRACT
Lariciresinol is a dietary lignan that accounts for a significant portion of the total phytoestrogen intake from Western foods. Recent epidemiological studies suggest that high dietary intake of lignans and lariciresinol is associated with reduced breast cancer risk. However, no causal relationship between lariciresinol intake and breast cancer development has been established. In this study, we investigated for the first time the effects and possible mechanisms of action of lariciresinol on hormone responsive mammary cancer in vivo in dimethylbenz[a]anthracene induced mammary cancer in rats, and in human MCF-7 breast cancer xenografts in athymic mice. For tumor bearing rats, lariciresinol (3 or 15 mg/kg of body weight) or vehicle was administered p.o. daily for 9 weeks. For E2-maintained ovariectomized athymic mice bearing orthotopic MCF-7 tumors, control diet (AIN-93G) or lariciresinol containing diet (AIN-93G supplemented with 20 or 100 mg of lariciresinol/kg of diet) was administered for 5 weeks. In both models, lariciresinol administration inhibited the tumor growth and tumor angiogenesis. In MCF-7 cells, enterolactone significantly inhibited the E2-stimulated VEGF secretion. Moreover, in MCF-7 xenografts, lariciresinol administration enhanced tumor cell apoptosis and increased estrogen receptor beta expression. Lariciresinol and its further metabolites secoisolariciresinol, enterodiol and enterolactone were found in serum of both rats and athymic mice confirming a similar lignan metabolism pattern as in humans. These findings indicate conceivable importance of dietary lignan lariciresinol in inhibition of breast cancer development.
Sigma-Aldrich
(+)-Lariciresinol, ≥95.0% (HPLC)
Pricing and availability is not currently available.