Merck
  • Silver pyridine-2-sulfonate complex - its characterization, DNA binding, topoisomerase I inhibition, antimicrobial and anticancer response.
Silver pyridine-2-sulfonate complex - its characterization, DNA binding, topoisomerase I inhibition, antimicrobial and anticancer response.
Journal of inorganic biochemistry (2018-07-02)
Michaela Rendošová, Zuzana Vargová, Danica Sabolová, Natália Imrichová, Daniela Hudecová, Róbert Gyepes, Boris Lakatoš, Katarína Elefantová
ABSTRACT
In the current study the ability of silver pyridine-2-sulfonate complex to exert multiple biological activities is compared with the pharmacological action of silver sulfadiazine (AgSD). Polymeric form of {[Ag(py-2-SO3)]}n (AgPS) was synthesized and characterized by analytical techniques (IR, CHN, TG/DTA, MS) and its molecular formula was established. The crystal structure was determined by X-ray diffraction method and the polymeric complex crystallizes in the triclinic P-1 space group. The stability of Ag(I) complex was verified by 1H and 13C NMR measurements and the interaction with calf thymus DNA through UV-VIS and fluorescence quenching experiments was studied. The Ag(I) complex was able to interact with DNA by dual binding mode: partial intercalation along groove binding. The binding constants were calculated to be in the order of 103 M-1. Topoisomerase I inhibition study have shown that silver complex is inhibiting its activity at concentration of 30 μM. The cytotoxic activity of AgPS and AgSD against mouse leukaemia L1210 S, R and T cell line was also evaluated. AgPS showed higher cytotoxicity than AgSD after 48 h incubation. The results suggest that mechanism of cell death is necrosis with a contribution of late apoptosis. Antimicrobial testing indicates higher growth inhibition effect of AgPS with comparison to commercially available AgSD.
Sigma-Aldrich
2-Pyridinesulfonic acid, 97%