Merck
  • A sensitive and efficient procedure for the high-throughput determination of nine urinary metabolites of pyrethroids by GC-MS/MS and its application in a sample of Japanese children.
A sensitive and efficient procedure for the high-throughput determination of nine urinary metabolites of pyrethroids by GC-MS/MS and its application in a sample of Japanese children.
Analytical and bioanalytical chemistry (2018-07-27)
Yuko Ueda, Masaya Oda, Isao Saito, Risa Hamada, Takaaki Kondo, Michihiro Kamijima, Jun Ueyama
ABSTRACT
Four pyrethroids (PYRs), metofluthrin, profluthrin, tefluthrin, and transfluthrin, which were newly developed and have relatively high vapor activity at ambient temperature, are now playing a key role in safely controlling insects in our daily lives. We developed a sensitive and high-throughput determination method for urinary metabolites derived from the newly developed PYR, e.g., 2,3,5,6-tetrafluoro-1,4-benzenedimethanol (HOCH2-FB-Al), 2,3,5,6-tetrafluorobenzyl alcohol (FB-Al), and other PYR metabolites such as trans-chrysanthemumdicarboxylic acid (trans-CDCA) and 3-phenoxybenzoic acid (3PBA). After high temperature acid hydrolysis of 2 mL urine sample in 24-deep well plate, the PYR metabolites were extracted by semi-automated liquid-liquid extraction with tert-butyl methyl ether. N,O-Bis (trimethylsilyl) trifluoroacetamide containing 1% trimethylchlorosilane or 1,1,1,3,3,3-hexafluoroisopropanol were used for the derivatization of PYR metabolites, and the derivatized metabolites were analyzed separately by GC-MS/MS equipped with dual injector system (DB-5MS and mid- to high-polarity phase Rtx-65 columns). The derivatization and evaporation conditions were mainly optimized for improving sensitivity and reproducibility. The mean within-run day precisions were less than 18.4% (relative standard deviation, %RSD) with low detection limits ranging from 0.01 μg/L for HOCH2-FB-Al to 0.06 μg/L for trans-CDCA. This method was successfully applied to urine samples obtained from 50 3-year-old children with high detection frequencies (e.g., 82% for HOCH2-FB-Al and 84% for FB-Al). This method may be a pivotal tool for developing risk assessment from PYR exposure in the general population.
Sigma-Aldrich
2-Phenoxybenzoic acid, 98%