Merck
  • Highly Selective Hydrogenation of Aromatic Ketones and Phenols Enabled by Cyclic (Amino)(alkyl)carbene Rhodium Complexes.
Highly Selective Hydrogenation of Aromatic Ketones and Phenols Enabled by Cyclic (Amino)(alkyl)carbene Rhodium Complexes.
Journal of the American Chemical Society (2015-07-15)
Yu Wei, Bin Rao, Xuefeng Cong, Xiaoming Zeng
ABSTRACT
Air-stable Rh complexes ligated by strongly σ-donating cyclic (amino)(alkyl)carbenes (CAACs) show unique catalytic activity for the selective hydrogenation of aromatic ketones and phenols by reducing the aryl groups. The use of CAAC ligands is essential for achieving high selectivity and conversion. This method is characterized by its good compatibility with unsaturated ketones, esters, carboxylic acids, amides, and amino acids and is scalable without detriment to its efficiency.