Merck
  • Gold nanoparticles conjugating recombinant influenza hemagglutinin trimers and flagellin enhanced mucosal cellular immunity.
Gold nanoparticles conjugating recombinant influenza hemagglutinin trimers and flagellin enhanced mucosal cellular immunity.
Nanomedicine : nanotechnology, biology, and medicine (2018-04-13)
Chao Wang, Wandi Zhu, Yuan Luo, Bao-Zhong Wang
ABSTRACT
The immunogenicity of subunit vaccines can be augmented by formulating them into nanoparticles. We conjugated recombinant trimetric influenza A/Aichi/2/68(H3N2) hemagglutinin (HA) onto functionalized gold nanoparticle (AuNP) surfaces in a repetitive, oriented configuration. To further improve the immunogenicity, we generated Toll-like receptor 5 (TLR5) agonist flagellin (FliC)-coupled AuNPs as particulate adjuvants. Intranasal immunizations with an AuNP-HA and AuNP-FliC particle mixture elicited strong mucosal and systemic immune responses that protected hosts against lethal influenza challenges. Compared with the AuNP-HA alone group, the addition of AuNP-FliC improved mucosal B cell responses as characterized by elevated influenza specific IgA and IgG levels in nasal, tracheal, and lung washes. AuNP-HA/AuNP-FliC also stimulated antigen-specific interferon-γ (IFN-γ)-secreting CD4+ cell proliferation and induced strong effector CD8+ T cell activation. Our results indicate that intranasal co-delivery of antigen and adjuvant-displaying AuNPs enhanced vaccine efficacy by inducing potent cellular immune responses.
Sigma-Aldrich
Propargyl ether, 98%