Merck
  • The anti-inflammatory effect of microRNA-383-3p interacting with IL1R2 against homocysteine-induced endothelial injury in rat coronary arteries.
The anti-inflammatory effect of microRNA-383-3p interacting with IL1R2 against homocysteine-induced endothelial injury in rat coronary arteries.
Journal of cellular biochemistry (2018-04-26)
Zheng Lian, Feng-Feng Lv, Jing Yu, Jia-Wang Wang
ABSTRACT
MicroRNAs (miRs) are widely reported to be novel biomarkers involved in the process of coronary atherosclerosis (CAS). Hence, this study aims to explore the function of miR-383-3p targeting IL1R2 on inflammatory injury of coronary artery endothelial cells (CAECs) in CAS. The underlying regulatory mechanisms of miR-383-3p were analyzed in concert with the treatment of miR-383-3p mimics, miR-383-3p inhibitors, and the combination of miR-383-3p inhibitors and siRNA against IL1R2 in homocysteine (HCY)-induced CAECs. MTT, Hoechst 33258 staining, and tube formation assay were employed in order to measure cell viability, apoptosis, and tube formation, respectively. The levels of IL-1β, IL-6, IL-10, and IL-18 were determined by ELISA. IL1R2 was verified as the target gene of miR-383-3p by dual-luciferase reporter gene assay. MiR-383-3p was down-regulated in myocardial tissues of AS rats while IL1R2 was the reciprocal. The up-regulation of miR-383-3p decreased the levels of IL1R2, caspase-1, IL-1β, IL-6, and IL-18 expressions, as well as cell apoptosis rate in the HCY-induced CAECs, while IL-10 expression, cell viability, and tube formation ability were increased. These results were contraindicated in the HCY-induced CAECs treated by miR-383-3p inhibitors. In conclusion, miR-383-3p mediating IL1R2 prevents HCY-induced apoptosis and inflammation injury in CAECs through the inhibition of the activation of inflammasome signaling pathway. These findings highly indicate that miR-383-3p may be beneficial in the prevention of CAS and other cardiovascular diseases.